missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetramethylbenzene, 97+%

Catalog No. p-7044091
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0449322 100 g
AAL0449336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0449322 Supplier Thermo Scientific Chemicals Supplier No. L0449322
Only null left

CAS: 95-93-2 | C10H14 | 134.222 g/mol

1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

Solubility
Soluble in alcohol, ether, benzene, and acetone. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 95-93-2
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008528
InChI Key SQNZJJAZBFDUTD-UHFFFAOYSA-N
Synonym durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren
PubChem CID 7269
ChEBI CHEBI:38978
IUPAC Name 1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C=C1C)C)C

Specifications

Melting Point 77°C to 80°C
Boiling Point 191°C to 193°C
Flash Point 73°C (163°F)
Quantity 100 g
UN Number UN1325
Beilstein 1903393
Merck Index 14,3468
Solubility Information Soluble in alcohol,ether,benzene,and acetone. Insoluble in water.
Formula Weight 134.22
Percent Purity ≥97%
Chemical Name or Material 1,2,4,5-Tetramethylbenzene
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 202-465-7
RTECSNumber DC0500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only