Learn More
1,2,4,5-Tetramethylbenzene, 97+%
CAS: 95-93-2 | C10H14 | 134.22 g/mol
$260.98 - $260.98
Chemical Identifiers
| CAS | 95-93-2 |
|---|---|
| Molecular Formula | C10H14 |
| Molecular Weight (g/mol) | 134.22 |
| InChI Key | SQNZJJAZBFDUTD-UHFFFAOYSA-N |
| Synonym | durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren |
| PubChem CID | 7269 |
| ChEBI | CHEBI:38978 |
| IUPAC Name | 1,2,4,5-tetramethylbenzene |
| SMILES | CC1=CC(=C(C=C1C)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC138375000
|
Thermo Scientific Chemicals
138375000 |
500 g |
Each for $260.98
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 95-93-2 | |
| 134.22 | |
| durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren | |
| CHEBI:38978 | |
| CC1=CC(=C(C=C1C)C)C |
| C10H14 | |
| SQNZJJAZBFDUTD-UHFFFAOYSA-N | |
| 7269 | |
| 1,2,4,5-tetramethylbenzene |
Specifications
| 95-93-2 | |
| 196.0°C to 197.0°C | |
| Authentic | |
| C10H14 | |
| 500 g | |
| 15,3515 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene | |
| CC1=CC(=C(C=C1C)C)C | |
| 134.22 | |
| CHEBI:38978 | |
| 97+% |
| 78.0°C to 82.0°C | |
| 73°C | |
| 97% min. (GC) | |
| C6H2(CH3)4 | |
| 05,431 | |
| durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren | |
| SQNZJJAZBFDUTD-UHFFFAOYSA-N | |
| 1,2,4,5-tetramethylbenzene | |
| 7269 | |
| 134.22 | |
| 1,2,4,5-Tetramethylbenzene |
Safety and Handling
GHS H Statement
Flammable solid.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
GHS Signal Word: Danger
EINECSNumber : 202-465-7
RUO – Research Use Only