missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Cyclohexanetetracarboxylic Dianhydride (purified by sublimation) 98.0+%, TCI America™

Catalog No. C29191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29191G 1 g
C29195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29191G Supplier TCI America Supplier No. C29191G
Only null left

Chemical Identifiers

CAS 2754-41-8
Molecular Formula C10H8O6
Molecular Weight (g/mol) 224.17
MDL Number MFCD13704912
InChI Key LJMPOXUWPWEILS-UHFFFAOYNA-N
PubChem CID 14453667
IUPAC Name 5,11-dioxatricyclo[7.3.0.0³,⁷]dodecane-4,6,10,12-tetrone
SMILES O=C1OC(=O)C2CC3C(CC12)C(=O)OC3=O

Specifications

Melting Point 240°C
Color White
Quantity 1 g
Formula Weight 224.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Cyclohexanetetracarboxylic Dianhydride (purified by sublimation)