missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Cyclohexanetetracarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7125329
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21985G 5 g
C219825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21985G Supplier TCI America Supplier No. C21985G
Only null left

Chemical Identifiers

CAS 15383-49-0
Molecular Formula C10H12O8
Molecular Weight (g/mol) 260.198
MDL Number MFCD00435556
InChI Key ZPAKUZKMGJJMAA-UHFFFAOYSA-N
PubChem CID 2834946
IUPAC Name cyclohexane-1,2,4,5-tetracarboxylic acid
SMILES C1C(C(CC(C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 271°C
Color White-Yellow
Quantity 5 g
Formula Weight 260.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Cyclohexanetetracarboxylic Acid