missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Triphenylguanidine 97.0+%, TCI America™

Catalog No. T131625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T131625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T131625G Supplier TCI America Supplier No. T131625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-01-9
Molecular Formula C19H17N3
Molecular Weight (g/mol) 287.366
MDL Number MFCD00020648
InChI Key FUPAJKKAHDLPAZ-UHFFFAOYSA-N
Synonym n,n',n-triphenylguanidine, guanidine, n,n',n-triphenyl, sym-triphenylguanidine, guanidine, triphenyl, 1,2,3-trifenylguanidin, unii-i64b170qfg, 1,2,3-trifenylguanidin czech, guanidine, 1,2,3-triphenyl, phenyl 2-phenyl-1-phenylamino-2-azavinyl amine, triphenylguanidin
PubChem CID 7539
IUPAC Name 1,2,3-triphenylguanidine
SMILES C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)NC3=CC=CC=C3

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 25 g
Formula Weight 287.37
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Triphenylguanidine
RTECSNumber MF6825000
TSCA Yes