missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Trichloropropene (cis- and trans- mixture) 95.0+%, TCI America™
$84.70 - $587.18
Chemical Identifiers
| CAS | 96-19-5 |
|---|---|
| Molecular Formula | C3H3Cl3 |
| Molecular Weight (g/mol) | 145.41 |
| MDL Number | MFCD00018931 |
| InChI Key | HIILBTHBHCLUER-IWQZZHSRSA-N |
| PubChem CID | 5370144 |
| IUPAC Name | (1Z)-1,2,3-trichloroprop-1-ene |
| SMILES | ClC\C(Cl)=C\Cl |
Chemical Identifiers
| 96-19-5 | |
| 145.41 | |
| HIILBTHBHCLUER-IWQZZHSRSA-N | |
| (1Z)-1,2,3-trichloroprop-1-ene |
| C3H3Cl3 | |
| MFCD00018931 | |
| 5370144 | |
| ClC\C(Cl)=C\Cl |
Specifications
| 96-19-5 | |
| 145°C | |
| MFCD00018931 | |
| 2810 | |
| ClC\C(Cl)=C\Cl | |
| 145.41 | |
| 145.41 | |
| Liquid |
| Yellow | |
| C3H3Cl3 | |
| 25 g | |
| HIILBTHBHCLUER-IWQZZHSRSA-N | |
| (1Z)-1,2,3-trichloroprop-1-ene | |
| 5370144 | |
| ≥95.0% (GC) | |
| 1,2,3-Trichloropropene (cis- and trans- mixture) |
Safety and Handling
RTECSNumber : UD2450000
TSCA : Yes