missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trichloropropane, 98+%

CAS: 96-18-4 | C3H5Cl3 | 147.42 g/mol

$77.84 - $195.95

Chemical Identifiers

CAS 96-18-4
Molecular Formula C3H5Cl3
Molecular Weight (g/mol) 147.42
MDL Number MFCD00000946
InChI Key CFXQEHVMCRXUSD-UHFFFAOYSA-N
Synonym trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
PubChem CID 7285
ChEBI CHEBI:34036
IUPAC Name 1,2,3-trichloropropane
SMILES ClCC(Cl)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0431230
View Documents
Thermo Scientific Chemicals
L0431230
250 g
Each for $77.84
Only null left
 
AAL043120B
View Documents
Thermo Scientific Chemicals
L043120B
1000 g
Each for $195.95
Only null left
 
Description

Description

1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.

Solubility
Slightly soluble in water (2 g/L at 25°C).

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Specifications

Chemical Identifiers

96-18-4
147.42
CFXQEHVMCRXUSD-UHFFFAOYSA-N
7285
1,2,3-trichloropropane
C3H5Cl3
MFCD00000946
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
CHEBI:34036
ClCC(Cl)CCl

Specifications

96-18-4
1.385
74°C (165°F)
1.4822
250 g
1732068
Slightly soluble in water (2g/L at 25°C).
ClCC(Cl)CCl
147.42
CHEBI:34036
≥98%
-14°C
156°C to 158°C
C3H5Cl3
MFCD00000946
UN2810
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
CFXQEHVMCRXUSD-UHFFFAOYSA-N
1,2,3-trichloropropane
7285
147.43
1,2,3-Trichloropropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H360-H302-H312-H332-H227
May cause cancer.
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

P201-P202-P210-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P370+P378q-P501c

H227-H302+H312-H350-H360F

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 202-486-1

RTECSNumber : TZ9275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only