missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trichloropropane, 98+%

Catalog No. AAL0431230
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0431230 250 g
AAL043120B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0431230 Supplier Thermo Scientific Chemicals Supplier No. L0431230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-18-4 | C3H5Cl3 | 147.42 g/mol

1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.

Solubility
Slightly soluble in water (2 g/L at 25°C).

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com

Chemical Identifiers

CAS 96-18-4
Molecular Formula C3H5Cl3
Molecular Weight (g/mol) 147.42
MDL Number MFCD00000946
InChI Key CFXQEHVMCRXUSD-UHFFFAOYSA-N
Synonym trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
PubChem CID 7285
ChEBI CHEBI:34036
IUPAC Name 1,2,3-trichloropropane
SMILES ClCC(Cl)CCl

Specifications

Melting Point -14°C
Density 1.385
Boiling Point 156°C to 158°C
Flash Point 74°C (165°F)
Refractive Index 1.4822
Quantity 250 g
UN Number UN2810
Beilstein 1732068
Solubility Information Slightly soluble in water (2g/L at 25°C).
Formula Weight 147.43
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trichloropropane
Show More Show Less
Hazard Category H227-H302+H312-H350-H360F
Hazard Statement GHS H Statement
H350-H360-H302-H312-H332-H227
May cause cancer.
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P201-P202-P210-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 202-486-1
RTECSNumber TZ9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only