missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trichlorobenzene 98.0+%, TCI America™

Catalog No. T037725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T037725G 25 g
T0377500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T037725G Supplier TCI America Supplier No. T037725G

Chemical Identifiers

CAS 87-61-6
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000537
InChI Key RELMFMZEBKVZJC-UHFFFAOYSA-N
Synonym trichlorobenzene, vic-trichlorobenzene, 1,2,6-trichlorobenzene, benzene, 1,2,3-trichloro, benzene, trichloro, unii-nur9777ik4, ccris 5944, dsstox_cid_6193, dsstox_rid_78052, dsstox_gsid_26193
PubChem CID 6895
ChEBI CHEBI:35289
IUPAC Name 1,2,3-trichlorobenzene
SMILES ClC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 53°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 3077
Boiling Point 219°C
Formula Weight 181.44
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,2,3-Trichlorobenzene
EINECSNumber (3)-0074
RTECSNumber DC2095000
TSCA Yes