missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 67°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 260.24 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose |
Safety and Handling
| TSCA | No |