missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose 98.0+%, TCI America™

Catalog No. p-7137313
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26075G 5 g
T260725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26075G Supplier TCI America Supplier No. T26075G
Only null left

Chemical Identifiers

CAS 62211-93-2
Molecular Formula C11H16O7
Molecular Weight (g/mol) 260.24
MDL Number MFCD08458459,MFCD08458459
InChI Key NXEJETQVUQAKTO-UHFFFAOYNA-N
Synonym 1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
PubChem CID 70847275
IUPAC Name 4,5-bis(acetyloxy)-2-methyloxolan-3-yl acetate
SMILES CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 5 g
Formula Weight 260.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose