missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose, 98%

Catalog No. AAJ6681803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6681803 1 g
AAJ6681806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6681803 Supplier Thermo Scientific Chemicals Supplier No. J6681803
Only null left

CAS: 62211-93-2 | C11H16O7 | 260.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62211-93-2
Molecular Formula C11H16O7
Molecular Weight (g/mol) 260.24
MDL Number MFCD08458459,MFCD08458459
InChI Key NXEJETQVUQAKTO-UHFFFAOYNA-N
Synonym 1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
PubChem CID 70847275
IUPAC Name [(2R,5S)-4,5-diacetyloxy-2-methyloxolan-3-yl] acetate
SMILES CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 65°C to 69°C
Color Yellow to Brown
Quantity 1 g
Solubility Information Soluble in ethanol
Formula Weight 260.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only