missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Thiadiazole-4-carboxylic acid, 97%

Catalog No. AAB2084203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2084203 1 g
AAB2084206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2084203 Supplier Thermo Scientific Chemicals Supplier No. B2084203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4100-13-4 | C3H2N2O2S | 130.121 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 4100-13-4
Molecular Formula C3H2N2O2S
Molecular Weight (g/mol) 130.121
MDL Number MFCD00052099
InChI Key HJZYBDPHAHGHAZ-UHFFFAOYSA-N
Synonym 1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid
PubChem CID 351418
IUPAC Name thiadiazole-4-carboxylic acid
SMILES C1=C(N=NS1)C(=O)O

Specifications

Melting Point 227°C to 228°C
Quantity 1 g
Beilstein 113707
Solubility Information Slightly soluble in water.
Formula Weight 130.12
Percent Purity 97%
Chemical Name or Material 1,2,3-Thiadiazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only