missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dihydrobenzo[b]furan-5-yl)ethan-1-one, 97%, Thermo Scientific™

Catalog No. KM05976DA
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM05976DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM05976DA Supplier Thermo Scientific Chemicals Supplier No. KM05976DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90843-31-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00068033
InChI Key MMVUJVASBDVNGJ-UHFFFAOYSA-N
Synonym 5-acetyl-2,3-dihydrobenzo b furan, 5-acetyl-2,3-dihydrobenzofuran, 1-2,3-dihydro-1-benzofuran-5-yl ethanone, 1-2,3-dihydrobenzofuran-5-yl ethanone, 1-2,3-dihydrobenzo b furan-5-yl ethan-1-one, 1-2,3-dihydro-benzofuran-5-yl-ethanone, 1-2,3-dihydro-5-benzofuranyl ethanone, 1-2,3-dihydro-1-benzofuran-5-yl ethan-1-one, 5-acetyl-2,3-dihydro-1-benzofuran
PubChem CID 145220
IUPAC Name 1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
SMILES CC(=O)C1=CC=C2OCCC2=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 1-(2, 3-Dihydrobenzo[b]furan-5-yl)ethan-1-one