missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Benzotriazole 98.0+%, TCI America™

Catalog No. p-7118348
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B009425G 25 g
B0094100G 100 g
B0094500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B009425G Supplier TCI America Supplier No. B009425G

Chemical Identifiers

CAS 95-14-7
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD00005699
InChI Key QRUDEWIWKLJBPS-UHFFFAOYSA-N
Synonym 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
PubChem CID 7220
ChEBI CHEBI:75331
IUPAC Name 2H-benzotriazole
SMILES C1=CC2=NNN=C2C=C1

Specifications

Melting Point 98°C
Color White-Yellow
pH 5
Boiling Point 204°C
Quantity 25 g
Formula Weight 119.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Benzotriazole
EINECSNumber (5)-0537
RTECSNumber DM1225000
TSCA Yes