missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetrahydropyridine, 98%, Thermo Scientific™

Catalog No. AC158870050
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158870050 5g Glass bottle
AC158870010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158870050 Supplier Thermo Scientific Chemicals Supplier No. 158870050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 694-05-3
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00005949
InChI Key FTAHXMZRJCZXDL-UHFFFAOYSA-N
Synonym pyridine, 1,2,3,6-tetrahydro, 3-piperideine, 1,2,5,6-tetrahydropyridine, unii-26rls9d255, delta 3-piperideine, .delta.3-piperidine, delta3-piperidine, acmc-1bcyc, 1,3,6-tetrahydropyridine, 1,5,6-tetrahydropyridine
PubChem CID 12750
ChEBI CHEBI:47860
IUPAC Name 1,2,3,6-tetrahydropyridine
SMILES C1CNCC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.9100g/mL
Boiling Point 108.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.48
Quantity 5g
Beilstein 20, III, 1912
Specific Gravity 0.91
Solubility Information Solubility in water: soluble.
Formula Weight 83.13
Percent Purity 98%
Chemical Name or Material 1,2,3,6-Tetrahydropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If eye irritation persists: Get medical advice/a
EINECSNumber 211-766-2

RUO – Research Use Only