missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6,7,8-Hexahydropyrene 98.0+%, TCI America™

Catalog No. H00861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00861G 1 g
H00865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H00861G Supplier TCI America Supplier No. H00861G
Only null left

Chemical Identifiers

CAS 1732-13-4
Molecular Formula C16H16
Molecular Weight (g/mol) 208.304
MDL Number MFCD00010584
InChI Key MBAIEZXRGAOPKH-UHFFFAOYSA-N
PubChem CID 74417
IUPAC Name 1,2,3,6,7,8-hexahydropyrene
SMILES C1CC2=C3C(=CC=C4C3=C(CCC4)C=C2)C1

Specifications

Melting Point 134°C
Color White
Quantity 1 g
Formula Weight 208.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,6,7,8-Hexahydropyrene