missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose, 98%

Catalog No. AAJ6607403
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6607403 1 g
AAJ6607406 5 g
AAJ6607409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6607403 Supplier Thermo Scientific Chemicals Supplier No. J6607403

CAS: 144490-03-9 | C13H18O9 | 318.278 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144490-03-9
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00674547
InChI Key IHNHAHWGVLXCCI-UMSGYPCISA-N
Synonym 1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate
PubChem CID 69431757
IUPAC Name [(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 80°C to 83°C
Color White
Quantity 1 g
Optical Rotation +12.7° (c=2.4 in Chloroform)
Formula Weight 318.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only