missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetraphenyl-1,3-cyclopentadiene 98.0+%, TCI America™

Catalog No. T13331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13331G 1 g
T13335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13331G Supplier TCI America Supplier No. T13331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15570-45-3
Molecular Formula C29H22
Molecular Weight (g/mol) 370.495
MDL Number MFCD00001355
InChI Key JCXLYAWYOTYWKM-UHFFFAOYSA-N
Synonym 1,2,3,4-tetraphenyl-1,3-cyclopentadiene, 1,2,3,4-tetraphenylcyclopentadiene, tetraphenylcyclopentadiene, 1,2,3,4-tetraphenylcyclopenta-1,3-diene, 1,2,3,4-tetraphebyl-1,3-cyclopentadene, 2,3,4-triphenylcyclopenta-1,3-dien-1-yl benzene, 2,4,5-triphenylcyclopenta-1,4-dien-1-yl benzene, benzene,1,1',1,1'-1,3-cyclopentadiene-1,2,3,4-tetrayl tetrakis, acmc-209dcl, 1,3-cyclopentadiene, 1,2,3,4-tetraphenyl
PubChem CID 84991
IUPAC Name (2,3,4-triphenylcyclopenta-1,3-dien-1-yl)benzene
SMILES C1C(=C(C(=C1C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 370.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetraphenyl-1,3-cyclopentadiene