missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetramethylbenzene, 95%

Catalog No. AAL1975606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1975606 5 g
AAL1975614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1975606 Supplier Thermo Scientific Chemicals Supplier No. L1975606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 488-23-3 | C10H14 | 134.222 g/mol

1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 488-23-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008521
InChI Key UOHMMEJUHBCKEE-UHFFFAOYSA-N
Synonym prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene
PubChem CID 10263
ChEBI CHEBI:38997
IUPAC Name 1,2,3,4-tetramethylbenzene
SMILES CC1=C(C(=C(C=C1)C)C)C

Specifications

Density 0.9
Boiling Point 203°C
Flash Point 68°C (154°F)
Refractive Index 1.519
Quantity 5 g
Beilstein 1904390
Solubility Information Immiscible with water.
Formula Weight 134.22
Percent Purity 95%
Chemical Name or Material 1,2,3,4-Tetramethylbenzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 207-673-1
RTECSNumber DC0465000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only