missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic Dianhydride, TCI America™

Supplier:  TCI America T26971G

Catalog No. T26971G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic Dianhydride
380°C
C12H12O6
GTDPSWPPOUPBNX-UHFFFAOYSA-N
252.22
64198-16-9
White-Yellow
1 g
252.22
Crystalline Powder

Chemical Identifiers

64198-16-9
C12H12O6
252.22
GTDPSWPPOUPBNX-UHFFFAOYSA-N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions