missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroquinaldine 97.0+%, TCI America™

Catalog No. T0112500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0112500G 500 g
T011225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0112500G Supplier TCI America Supplier No. T0112500G

Chemical Identifiers

CAS 1780-19-4
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00023886
InChI Key JZICUKPOZUKZLL-UHFFFAOYNA-N
Synonym 2-Methyl-1,2,3,4-tetrahydroquinoline, 1,2,3,4-Tetrahydro-2-methylquinoline
PubChem CID 96289
IUPAC Name 2-methyl-1,2,3,4-tetrahydroquinoline
SMILES CC1CCC2=CC=CC=C2N1

Specifications

Color Yellow
Boiling Point 250°C
Quantity 500 g
Formula Weight 147.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydroquinaldine