missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroisoquinoline 95.0+%, TCI America™

Catalog No. T1676500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1676500G 500 g
T167625G 25 g
T1676100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1676500G Supplier TCI America Supplier No. T1676500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-21-4
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00006896
InChI Key UWYZHKAOTLEWKK-UHFFFAOYSA-N
Synonym tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro isoquinoline, 1,2,3,4-tetrahydro-2-isoquinoline, 1,2,3,4-tetrahydro-2-azanaphthalene, unii-56w89fbx3e, chembl14346, 3,4-dihydro-1h-isoquinoline, tetrahydroisoquinolin
PubChem CID 7046
IUPAC Name 1,2,3,4-tetrahydroisoquinoline
SMILES C1CC2=CC=CC=C2CN1

Specifications

Melting Point -30°C
Color Yellow
Boiling Point 233°C
Quantity 500 g
Formula Weight 133.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline
RTECSNumber NX4900000
TSCA Yes