missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydrocyclopenta[b]indole 97.0+%, TCI America™
$171.49 - $171.49
Chemical Identifiers
| CAS | 2047-91-8 |
|---|---|
| Molecular Formula | C11H11N |
| Molecular Weight (g/mol) | 157.216 |
| MDL Number | MFCD00225387 |
| InChI Key | HZDXFZHFEASSBM-UHFFFAOYSA-N |
| PubChem CID | 270305 |
| IUPAC Name | 1,2,3,4-tetrahydrocyclopenta[b]indole |
| SMILES | C1CC2=C(C1)NC3=CC=CC=C23 |
Chemical Identifiers
| 2047-91-8 | |
| 157.216 | |
| HZDXFZHFEASSBM-UHFFFAOYSA-N | |
| 1,2,3,4-tetrahydrocyclopenta[b]indole |
| C11H11N | |
| MFCD00225387 | |
| 270305 | |
| C1CC2=C(C1)NC3=CC=CC=C23 |
Specifications
| 2047-91-8 | |
| C11H11N | |
| 5 g | |
| C1CC2=C(C1)NC3=CC=CC=C23 | |
| 157.216 | |
| 157.22 | |
| Crystalline Powder |
| 105°C | |
| MFCD00225387 | |
| HZDXFZHFEASSBM-UHFFFAOYSA-N | |
| 1,2,3,4-tetrahydrocyclopenta[b]indole | |
| 270305 | |
| ≥97.0% (GC) | |
| 1,2,3,4-Tetrahydrocyclopenta[b]indole |
Safety and Handling
TSCA : No