missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydro-9-methylcarbazol-4-one 98.0+%, TCI America™
$57.79 - $264.70
Chemical Identifiers
| CAS | 27387-31-1 |
|---|---|
| Molecular Formula | C13H13NO |
| Molecular Weight (g/mol) | 199.253 |
| MDL Number | MFCD00173748 |
| InChI Key | HHJUJCWZKJMCLC-UHFFFAOYSA-N |
| Synonym | 1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole |
| PubChem CID | 598875 |
| IUPAC Name | 9-methyl-2,3-dihydro-1H-carbazol-4-one |
| SMILES | CN1C2=C(C(=O)CCC2)C3=CC=CC=C31 |
Chemical Identifiers
| 27387-31-1 | |
| 199.253 | |
| HHJUJCWZKJMCLC-UHFFFAOYSA-N | |
| 598875 | |
| CN1C2=C(C(=O)CCC2)C3=CC=CC=C31 |
| C13H13NO | |
| MFCD00173748 | |
| 1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole | |
| 9-methyl-2,3-dihydro-1H-carbazol-4-one |
Specifications
| 27387-31-1 | |
| White-Yellow | |
| MFCD00173748 | |
| 1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole | |
| CN1C2=C(C(=O)CCC2)C3=CC=CC=C31 | |
| 199.253 | |
| 199.25 | |
| Crystalline Powder |
| 201°C | |
| C13H13NO | |
| 5 g | |
| HHJUJCWZKJMCLC-UHFFFAOYSA-N | |
| 9-methyl-2,3-dihydro-1H-carbazol-4-one | |
| 598875 | |
| ≥98.0% (GC) | |
| 1,2,3,4-Tetrahydro-9-methylcarbazol-4-one |
Safety and Handling
TSCA : No