missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose, 98%

Catalog No. p-7043219
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66307ME 500 mg
AAJ6630703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAJ66307ME missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' J66307ME
Only null left

CAS: 13100-46-4 | C14H20O10 | 348.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13100-46-4
Molecular Formula C14H20O10
Molecular Weight (g/mol) 348.30
MDL Number MFCD00063260
InChI Key FEQXFAYSNRWXDW-UHFFFAOYNA-N
Synonym 1,2,3,4-tetra-o-acetyl-beta-d-glucopyranose, beta-d-glucose 1,2,3,4-tetraacetate, 2s,3r,4s,5r,6r-6-hydroxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-hydroxymethyl oxan-3-yl acetate, 1,2,3,4-tetra-o-acetylglucopyranose, beta-d-glucopyranose 1,2,3,4-tetraacetate, 1,2,3,4-tetra-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose, 1,2,3,4-tetraacetate, b-d-glucopyranose,1,2,3,4-tetraacetate
PubChem CID 83136
IUPAC Name [(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(hydroxymethyl)oxan-3-yl] acetate
SMILES CC(=O)OC1OC(CO)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 126°C to 128°C
Color White
Quantity 500 mg
Solubility Information Soluble in chloroform
Optical Rotation +8° (c=6 in Chloroform)
Formula Weight 348.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose
EINECSNumber 236-018-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only