missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Cyclopentanetetracarboxylic Dianhydride 98.0+%, TCI America™

Catalog No. C08575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08575G 5 g
C085725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08575G Supplier TCI America Supplier No. C08575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6053-68-5 | C9H6O6 | 210.141 g/mol

Chemical Identifiers

CAS 6053-68-5
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.141
MDL Number MFCD00046872
InChI Key NLWBEORDOPDUPM-UHFFFAOYSA-N
PubChem CID 110757
SMILES C1C2C(C3C1C(=O)OC3=O)C(=O)OC2=O

Specifications

Melting Point 213°C
Color White
Quantity 5 g
Formula Weight 210.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Cyclopentanetetracarboxylic Dianhydride