missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Cyclopentanetetracarboxylic Dianhydride (purified by sublimation) 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 216°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 210.14 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2,3,4-Cyclopentanetetracarboxylic Dianhydride (purified by sublimation) |
Safety and Handling
| TSCA | Yes |