missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Butanetetracarboxylic acid, 99+%

CAS: 1703-58-8 | C8H10O8 | 234.16 g/mol

$138.45 - $362.29

Chemical Identifiers

CAS 1703-58-8
Molecular Formula C8H10O8
Molecular Weight (g/mol) 234.16
MDL Number MFCD00002722
InChI Key GGAUUQHSCNMCAU-UHFFFAOYSA-N
Synonym 1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa
PubChem CID 15560
IUPAC Name butane-1,2,3,4-tetracarboxylic acid
SMILES C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366761000
SDS View Documents
Thermo Scientific Chemicals
366761000
100 g Glass bottle N/A
Only null left
 
AC366765000
SDS View Documents
Thermo Scientific Chemicals
366765000
500 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1703-58-8
234.16
GGAUUQHSCNMCAU-UHFFFAOYSA-N
15560
C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O
C8H10O8
MFCD00002722
1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa
butane-1,2,3,4-tetracarboxylic acid

Specifications

1703-58-8
100.0
White
99+%
C8H10O8
100 g
Solubility in water: soluble. Other solubilities: soluble in ethanol
C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O
234.16
234.16
Crystalline Powder
97.5
195.0°C to 197.0°C
Authentic
Glass bottle
MFCD00002722
1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa
GGAUUQHSCNMCAU-UHFFFAOYSA-N
butane-1,2,3,4-tetracarboxylic acid
15560
99+%
1,2,3,4-Butanetetracarboxylic acid, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

EINECSNumber : 216-938-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only