Learn More
1,2,3,4-Butanetetracarboxylic acid, 98+%
CAS: 1703-58-8 | C8H10O8 | 234.16 g/mol
$119.88 - $365.42
Chemical Identifiers
| CAS | 1703-58-8 |
|---|---|
| Molecular Formula | C8H10O8 |
| Molecular Weight (g/mol) | 234.16 |
| MDL Number | MFCD00002722 |
| InChI Key | GGAUUQHSCNMCAU-UHFFFAOYSA-N |
| Synonym | 1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa |
| PubChem CID | 15560 |
| IUPAC Name | butane-1,2,3,4-tetracarboxylic acid |
| SMILES | C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1661222
|
Thermo Scientific Chemicals
A1661222 |
100 g |
Each for $119.88
|
|
|||||
|
AAA1661236
|
Thermo Scientific Chemicals
A1661236 |
500 g |
Each for $365.42
|
|
|||||
Description
1,2,3,4-Butanetetracarboxylic acid is used as a cross linking agent to prepare cotton cellulose in order to improve anti-prilling and flame retardant properties. It is used as a spacer in the cross-linking of titania nanoparticles to cotton. Further, it is used as a nonformaldehyde durable press finishing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,3,4-Butanetetracarboxylic acid is used as a cross linking agent to prepare cotton cellulose in order to improve anti-prilling and flame retardant properties. It is used as a spacer in the cross-linking of titania nanoparticles to cotton. Further, it is used as a nonformaldehyde durable press finishing agent.
Solubility
Soluble in water and ethanol.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 1703-58-8 | |
| 234.16 | |
| GGAUUQHSCNMCAU-UHFFFAOYSA-N | |
| 15560 | |
| C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O |
| C8H10O8 | |
| MFCD00002722 | |
| 1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa | |
| butane-1,2,3,4-tetracarboxylic acid |
Specifications
| 1703-58-8 | |
| C8H10O8 | |
| 100 g | |
| 1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa | |
| GGAUUQHSCNMCAU-UHFFFAOYSA-N | |
| butane-1,2,3,4-tetracarboxylic acid | |
| 15560 | |
| ≥98% |
| ∼190°C to 195°C (decomposition) | |
| MFCD00002722 | |
| 1729167 | |
| Soluble in water and ethanol. | |
| C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O | |
| 234.16 | |
| 234.16 | |
| 1,2,3,4-Butanetetracarboxylic acid |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
EINECSNumber : 216-938-0
RTECSNumber : EK6100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only