missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine Copper(II) (purified by sublimation) 98.0+%, TCI America™

Catalog No. H11941G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11941G 1 g
H1194100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11941G Supplier TCI America Supplier No. H11941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14916-87-1
Molecular Formula C32H2CuF16N8
Molecular Weight (g/mol) 865.944
MDL Number MFCD00274642
InChI Key NKBVANSQKHVZHF-UHFFFAOYSA-N
Synonym Copper(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine, F16CuPc
PubChem CID 87077853
SMILES C12=C(C(=C(C(=C1F)F)F)F)C3=NC4=NC(=NC5=NC(=NC6=C7C(=C(N6)N=C2N3)C(=C(C(=C7F)F)F)F)C8=C5C(=C(C(=C8F)F)F)F)C9=C4C(=C(C(=C9F)F)F)F.[Cu]

Specifications

Color Purple-Red
Quantity 1 g
Formula Weight 863.93
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine Copper(II) (purified by subli