missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose 97.0+%, TCI America™

Catalog No. P15145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P15145G Supplier TCI America Supplier No. P15145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25941-03-1
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD05864874
InChI Key LPTITAGPBXDDGR-WHWZVRATSA-N
Synonym 1,2,3,4,6-penta-o-acetyl-d-mannopyranose, d-mannopyranose pentaacetate, 3s,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 2r,3r,4s,5s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, mannopyranose pentaacetate, d-mannopyranose, pentaacetate, 1,2,3,4,6-pentaacetate d-mannopyranose, d-mannopyranose,1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-dmannopyranose, 1,2,3,4,6-penta-o-acetyl-alpha,beta-d-mannose
PubChem CID 11811209
IUPAC Name [(2R,3R,4S,5S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 68°C
Color White
Quantity 5 g
Formula Weight 390.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
TSCA No
Recommended Storage Refrigerator