missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%

Catalog No. AAJ6697306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6697306 5 g
AAJ6697314 25 g
AAJ6697322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6697306 Supplier Thermo Scientific Chemicals Supplier No. J6697306

CAS: 25941-03-1 | C16H22O11 | 390.341 g/mol

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

Solubility
Soluble in dichloromethane, ether, ethyl acetate, methanol, and chloroform.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place. Store at -20°C.

Chemical Identifiers

CAS 25941-03-1
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD05864874
InChI Key LPTITAGPBXDDGR-WHWZVRATSA-N
Synonym 1,2,3,4,6-penta-o-acetyl-d-mannopyranose, d-mannopyranose pentaacetate, 3s,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 2r,3r,4s,5s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, mannopyranose pentaacetate, d-mannopyranose, pentaacetate, 1,2,3,4,6-pentaacetate d-mannopyranose, d-mannopyranose,1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-dmannopyranose, 1,2,3,4,6-penta-o-acetyl-alpha,beta-d-mannose
PubChem CID 11811209
IUPAC Name [(2R,3R,4S,5S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 66°C to 72°C
Color White
Quantity 5 g
Solubility Information Soluble in dichloromethane,ether,ethyl acetate,methanol,and chloroform.
Optical Rotation +54° (c=1 in Chloroform)
Formula Weight 390.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only