missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%

Catalog No. p-7043263
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAJ6668506 5 g
AAJ6668514 25 g
2 options

Catalog No. AAJ6668506

Supplier: Thermo Scientific Chemicals J6668506

Only null left

CAS: 4163-65-9 | C16H22O11 | 390.341 g/mol

α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4163-65-9
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00069798
InChI Key LPTITAGPBXDDGR-OWYFMNJBSA-N
Synonym a-d-mannose pentaacetate, alpha-d-mannose pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-mannopyranose, alpha-d-mannopyranose pentaacetate, 1-o,2-o,3-o,4-o,6-o-pentaacetyl-a-d-mannopyranose, man oac 5, a-d-mannosepentaacetate, pubchem22277, 1,2,3,4,6-penta-o-acetyl-, a-d-mannopyranose, 1,2,3,4,6-penta-o-acetyl-a-d-mannopyranose
PubChem CID 11741089
IUPAC Name [(2R,3R,4S,5S,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in chloroform
Optical Rotation +54° (c=1 in Chloroform)
Formula Weight 390.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only