missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene 98.0+%, TCI America™

Catalog No. P2297100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2297100MG 100 mg
P2297500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2297100MG Supplier TCI America Supplier No. P2297100MG
Only null left

Chemical Identifiers

CAS 312959-24-3
Molecular Formula C48H47FeP
Molecular Weight (g/mol) 710.72
MDL Number MFCD03788940
Synonym QPhos
IUPAC Name 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene
SMILES [Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 220°C
Quantity 100 mg
Formula Weight 710.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,5-Pentaphenyl-1′-(di-tert-butylphosphino)ferrocene