missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene 98.0+%, TCI America™
$42.87 - $42.87
Chemical Identifiers
| CAS | 2519-10-0 |
|---|---|
| Molecular Formula | C35H26 |
| Molecular Weight (g/mol) | 446.593 |
| InChI Key | YGLVWOUNCXBPJF-UHFFFAOYSA-N |
| PubChem CID | 635023 |
| IUPAC Name | (2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl)benzene |
| SMILES | C1=CC=C(C=C1)C2C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1633100MG
|
TCI America
P1633100MG |
100 mg |
Each for $42.87
|
|
|||||
Chemical Identifiers
| 2519-10-0 | |
| 446.593 | |
| 635023 | |
| C1=CC=C(C=C1)C2C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6 |
| C35H26 | |
| YGLVWOUNCXBPJF-UHFFFAOYSA-N | |
| (2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl)benzene |
Specifications
| 2519-10-0 | |
| C35H26 | |
| 1,2,3,4,5-pentaphenyl-1,3-cyclopentadiene, pentaphenylcyclopentadiene, cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, 2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl benzene, benzene,1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-yl benzene, acmc-1cek1, 2,3,4,5-tetraphenyl-1-cyclopenta-1,4-dienyl benzene, benzene, 1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dienyl benzene | |
| C1=CC=C(C=C1)C2C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6 | |
| 446.593 | |
| 446.59 | |
| Crystalline Powder |
| 256°C | |
| 100 mg | |
| YGLVWOUNCXBPJF-UHFFFAOYSA-N | |
| (2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl)benzene | |
| 635023 | |
| ≥98.0% (GC) | |
| 1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene |
Safety and Handling
TSCA : No