missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 256°C |
| Quantity | 100 mg |
| Synonym | 1,2,3,4,5-pentaphenyl-1,3-cyclopentadiene, pentaphenylcyclopentadiene, cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, 2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl benzene, benzene,1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-yl benzene, acmc-1cek1, 2,3,4,5-tetraphenyl-1-cyclopenta-1,4-dienyl benzene, benzene, 1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dienyl benzene |
| Formula Weight | 446.59 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene |
Safety and Handling
| TSCA | No |