missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5,6-Hexabromocyclohexane, Thermo Scientific Chemicals

Catalog No. p-7041961
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6085303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6085303 Supplier Thermo Scientific Chemicals Supplier No. J6085303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation

Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1837-91-8
Molecular Formula C6H6Br6
Molecular Weight (g/mol) 557.54
MDL Number MFCD00059127
InChI Key QFQZKISCBJKVHI-UHFFFAOYSA-N
Synonym benzene hexabromide, cyclohexane, 1,2,3,4,5,6-hexabromo, jak2 inhibitor ii, acmc-1bqjt, trans-alpha-benzene hexabromide, cyclohexane,2,3,4,5,6-hexabromo, 1,2,3,4,5,6-hexabromo-cyclohexane, #, 1,2,3,4,5,6-hexakis bromanyl cyclohexane, 1,2,3,4,5,6-hexabromocyclohexane hplc
PubChem CID 74603
IUPAC Name 1,2,3,4,5,6-hexabromocyclohexane
SMILES BrC1C(Br)C(Br)C(Br)C(Br)C1Br

Specifications

Color White
Quantity 1 g
Formula Weight 557.5
Physical Form Solid
Chemical Name or Material 1,2,3,4,5,6-Hexabromocyclohexane
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only