missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[2-(2-Hydroxyethoxy)ethyl]piperazine 98.0+%, TCI America™
$67.11 - $195.73
Chemical Identifiers
| CAS | 13349-82-1 |
|---|---|
| Molecular Formula | C8H18N2O2 |
| Molecular Weight (g/mol) | 174.244 |
| MDL Number | MFCD00074931 |
| InChI Key | FLNQAPQQAZVRDA-UHFFFAOYSA-N |
| Synonym | 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy |
| PubChem CID | 139436 |
| IUPAC Name | 2-(2-piperazin-1-ylethoxy)ethanol |
| SMILES | C1CN(CCN1)CCOCCO |
Chemical Identifiers
| 13349-82-1 | |
| 174.244 | |
| FLNQAPQQAZVRDA-UHFFFAOYSA-N | |
| 139436 | |
| C1CN(CCN1)CCOCCO |
| C8H18N2O2 | |
| MFCD00074931 | |
| 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy | |
| 2-(2-piperazin-1-ylethoxy)ethanol |
Specifications
| 13349-82-1 | |
| 172°C | |
| MFCD00074931 | |
| 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy | |
| C1CN(CCN1)CCOCCO | |
| 174.244 | |
| 174.24 | |
| Liquid |
| Yellow | |
| C8H18N2O2 | |
| 5 g | |
| FLNQAPQQAZVRDA-UHFFFAOYSA-N | |
| 2-(2-piperazin-1-ylethoxy)ethanol | |
| 139436 | |
| ≥98.0% (GC,T) | |
| 1-[2-(2-Hydroxyethoxy)ethyl]piperazine |
Safety and Handling
EINECSNumber : (5)-3830
TSCA : No