missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(2-Hydroxyethoxy)ethyl]piperazine 98.0+%, TCI America™

Catalog No. H11925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11925G 5 g
H119225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11925G Supplier TCI America Supplier No. H11925G

Chemical Identifiers

CAS 13349-82-1
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD00074931
InChI Key FLNQAPQQAZVRDA-UHFFFAOYSA-N
Synonym 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy
PubChem CID 139436
IUPAC Name 2-(2-piperazin-1-ylethoxy)ethanol
SMILES C1CN(CCN1)CCOCCO

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 174.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-[2-(2-Hydroxyethoxy)ethyl]piperazine
EINECSNumber (5)-3830
TSCA No