missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,6,6-Pentamethylpiperidine, 95%

Catalog No. AC416960050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416960050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC416960050 Supplier Thermo Scientific Chemicals Supplier No. 416960050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79-55-0 | C10H21N | 155.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-55-0
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.28
MDL Number MFCD00006493
InChI Key XULIXFLCVXWHRF-UHFFFAOYSA-N
Synonym pempidine, pyrilene, perolysen, tenormal, piperidine, 1,2,2,6,6-pentamethyl, pempidina dcit, pempidine inn:ban, pempidinum inn-latin, unii-n5i18ji9d6, 1,2,2,6,6-pentamethyl-piperidine
PubChem CID 6603
IUPAC Name 1,2,2,6,6-pentamethylpiperidine
SMILES CC1(CCCC(N1C)(C)C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8580g/mL
Boiling Point 187.0°C to 188.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.4600
Quantity 5 g
Specific Gravity 0.858
Merck Index 15, 7189
Formula Weight 155.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1,2,2,6,6-Pentamethylpiperidine, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/spark
EINECSNumber 201-211-2

RUO – Research Use Only