missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,6,6-Pentamethyl-4-piperidyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. p-7135166
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P151325G 25 g
P1513500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P151325G Supplier TCI America Supplier No. P151325G
Only null left

Chemical Identifiers

CAS 68548-08-3
Molecular Formula C14H25NO2
Molecular Weight (g/mol) 239.36
MDL Number MFCD08276414
InChI Key NWPIOULNZLJZHU-UHFFFAOYSA-N
Synonym Methacrylic Acid 1,2,2,6,6-Pentamethyl-4-piperidyl Ester
PubChem CID 10933588
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-yl 2-methylprop-2-enoate
SMILES CN1C(C)(C)CC(CC1(C)C)OC(=O)C(C)=C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 239.36
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,2,6,6-Pentamethyl-4-piperidyl Methacrylate (stabilized with MEHQ)
EINECSNumber (5)-6023
TSCA Yes
Recommended Storage Refrigerator