missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,2,6,6-Pentamethyl-4-piperidyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 239.36 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,2,2,6,6-Pentamethyl-4-piperidyl Methacrylate (stabilized with MEHQ) |
Sécurité et manipulation
| EINECSNumber | (5)-6023 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |