missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,6,6-Pentamethyl-4-piperidinol, 99%

Catalog No. AC300860050
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC300860050 5 g Glass bottle
AC300862500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC300860050 Supplier Thermo Scientific Chemicals Supplier No. 300860050
Only null left

CAS: 2403-89-6 | C10H21NO | 171.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2403-89-6
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.28
MDL Number MFCD00191220
InChI Key NWHNXXMYEICZAT-UHFFFAOYSA-N
Synonym 1,2,2,6,6-pentamethyl-4-piperidinol, 4-piperidinol, 1,2,2,6,6-pentamethyl, 4-hydroxy-1,2,2,6,6-pentamethylpiperidine, unii-lr520bam5v, 1,2,2,6,6-pentamethyl-4-hydroxypiperidine, lr520bam5v, methyl-taa-ol, hopemp, pubchem22251, acmc-209g8a
PubChem CID 75472
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-ol
SMILES CC1(CC(CC(N1C)(C)C)O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 76°C
Color Yellow
Density 0.9670g/mL
Boiling Point 238°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.967
Solubility Information Solubility in water: in water: 4.8% (20°C). Other solubilities: in ethanol: 55% (20°C),in acetone: 38% (20°C),in chloroform: 52% (20°C)
Viscosity 15.3 mPa.s (80°C)
Formula Weight 171.28
Percent Purity 99%
Physical Form Flakes
Chemical Name or Material 1,2,2,6,6-Pentamethyl-4-piperidinol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-292-8
TSCA TSCA

RUO – Research Use Only