missing translation for 'onlineSavingsMsg'
Learn More

1,18-Octadecanedicarboxylic acid

Catalog No. AA4200430
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4200430 250 g
AA4200404 2 g
AA4200409 10 g
AA4200418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4200430 Supplier Thermo Scientific Chemicals Supplier No. 04200430

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2424-92-2 | C20H38O4 | 342.52 g/mol

1,18-Octadecanedicarboxylic acid is used as an intermediate in organic synthesis, as dispersing agent, emulsifying agent, as lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2424-92-2
Molecular Formula C20H38O4
Molecular Weight (g/mol) 342.52
MDL Number MFCD00059627
InChI Key JJOJFIHJIRWASH-UHFFFAOYSA-N
Synonym eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
PubChem CID 75502
ChEBI CHEBI:73728
IUPAC Name icosanedioic acid
SMILES C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O

Specifications

Melting Point 127°C
Linear Formula HO2C(CH2)18CO2H
Quantity 250 g
Solubility Information Slightly soluble in water.
Formula Weight 342.52
Physical Form Powder
Chemical Name or Material 1,18-Octadecanedicarboxylic acid
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only