missing translation for 'onlineSavingsMsg'
Learn More

1,16-Hexadecanediol 95.0+%, TCI America™

Catalog No. p-7130603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05521G 1 g
H05525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05521G Supplier TCI America Supplier No. H05521G
Only null left

Chemical Identifiers

CAS 7735-42-4
Molecular Formula C16H34O2
Molecular Weight (g/mol) 258.45
MDL Number MFCD00002821
InChI Key GJBXIPOYHVMPQJ-UHFFFAOYSA-N
Synonym 1,16-Dihydroxyhexadecane, Hexadecamethylene Glycol
PubChem CID 82184
IUPAC Name hexadecane-1,16-diol
SMILES OCCCCCCCCCCCCCCCCO

Specifications

Melting Point 92°C
Color White
Boiling Point 200°C
Quantity 1 g
Formula Weight 258.45
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,16-Hexadecanediol
EINECSNumber (2)-0240
TSCA Yes