missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,12-Dodecanediol 99.0+%, TCI America™
$68.97 - $592.76
Chemical Identifiers
| CAS | 5675-51-4 |
|---|---|
| Molecular Formula | C12H26O2 |
| Molecular Weight (g/mol) | 202.34 |
| MDL Number | MFCD00004755 |
| InChI Key | GHLKSLMMWAKNBM-UHFFFAOYSA-N |
| Synonym | 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h |
| PubChem CID | 79758 |
| IUPAC Name | dodecane-1,12-diol |
| SMILES | OCCCCCCCCCCCCO |
Chemical Identifiers
| 5675-51-4 | |
| 202.34 | |
| GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
| 79758 | |
| OCCCCCCCCCCCCO |
| C12H26O2 | |
| MFCD00004755 | |
| 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
| dodecane-1,12-diol |
Specifications
| 5675-51-4 | |
| White | |
| C12H26O2 | |
| 25 g | |
| GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
| dodecane-1,12-diol | |
| 79758 | |
| ≥99.0% (GC) | |
| 1,12-Dodecanediol |
| 82°C | |
| 189°C | |
| MFCD00004755 | |
| 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
| OCCCCCCCCCCCCO | |
| 202.34 | |
| 202.34 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : Yes