missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dodecanediol Dimethacrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. D517225G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D517225G 25 g
D5172100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D517225G Supplier TCI America Supplier No. D517225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72829-09-5
Molecular Formula C20H34O4
Molecular Weight (g/mol) 338.488
MDL Number MFCD00080499
InChI Key HYQASEVIBPSPMK-UHFFFAOYSA-N
Synonym 1,12-Bis(methacryloyloxy)dodecane
PubChem CID 175191
IUPAC Name 12-(2-methylprop-2-enoyloxy)dodecyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Color Yellow
pH <7
Quantity 25 g
Formula Weight 338.49
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,12-Dodecanediol Dimethacrylate (stabilized with MEHQ)