missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,12-Dodecanediol Dimethacrylate (stabilized with MEHQ) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| pH | <7 |
| Quantity | 100 g |
| Formula Weight | 338.49 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,12-Dodecanediol Dimethacrylate (stabilized with MEHQ) |
Safety and Handling
| TSCA | Yes |