Learn More
1,12-Dodecanediol, 99%
CAS: 5675-51-4 | C12H26O2 | 202.34 g/mol
$108.56 - $247.14
Chemical Identifiers
| CAS | 5675-51-4 |
|---|---|
| Molecular Formula | C12H26O2 |
| Molecular Weight (g/mol) | 202.34 |
| MDL Number | MFCD00004755 |
| InChI Key | GHLKSLMMWAKNBM-UHFFFAOYSA-N |
| Synonym | 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h |
| PubChem CID | 79758 |
| IUPAC Name | dodecane-1,12-diol |
| SMILES | OCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2288514
|
Thermo Scientific Chemicals
B2288514 |
25 g |
Each for $108.56
|
|
|||||
|
AAB2288522
|
Thermo Scientific Chemicals
B2288522 |
100 g |
Each for $247.14
|
|
|||||
Description
1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.
Solubility
Soluble in alcohol and warm ether. Insoluble in water and petroleum ether.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 5675-51-4 | |
| 202.34 | |
| GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
| 79758 | |
| OCCCCCCCCCCCCO |
| C12H26O2 | |
| MFCD00004755 | |
| 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
| dodecane-1,12-diol |
Specifications
| 5675-51-4 | |
| 188°C to 190°C (12 mmHg) | |
| MFCD00004755 | |
| 1742760 | |
| Soluble in alcohol and warm ether. Insoluble in water and petroleum ether. | |
| OCCCCCCCCCCCCO | |
| 202.34 | |
| 202.34 | |
| 1,12-Dodecanediol |
| 81°C to 84°C | |
| C12H26O2 | |
| 25 g | |
| 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h | |
| GHLKSLMMWAKNBM-UHFFFAOYSA-N | |
| dodecane-1,12-diol | |
| 79758 | |
| 99% |
Safety and Handling
EINECSNumber : 227-133-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only